Läs mer
methyl 8,8-dioxo-8lambda6-thia-2,3,5,7-tetraazatricyclo[7.4.0.0,2,6]trideca-1(13),3,5,9,11-pentaene-4-carboxylate, TRC
Produktkod. 30477408
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30477408 10 mg
30483614 50 mg
30466630 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30477408 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM33582510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1311315-62-4
Molekylformel C10H8N4O4S
Molekylvikt (g/mol) 280.25
IUPAC-namn methyl 5,5-dioxo-1H-[1,2,4]triazolo[5,1-c][1,2,4]benzothiadiazine-2-carboxylate
LEDER COC(=O)C1=NC2=NS(=O)(=O)c3ccccc3N2N1
Specifikationer
Kemiskt namn eller material methyl 8,8-dioxo-8lambda6-thia-2,3,5,7-tetraazatricyclo[7.4.0.0,2,6]trideca-1(13),3,5,9,11-pentaene-4-carboxylate
Kvantitet 10 mg
InChI formel InChI=1S/C10H8N4O4S/c1-18-9(15)8-11-10-13-19(16,17)7-5-3-2-4-6(7)14(10)12-8/h2-5H,1H3,(H,11,12,13)
Formel vikt 280.0266