Läs mer
Methyl 9-Octadecenoate, TRC
Produktkod. 30179594
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179594 25 mg
30179024 250 mg
30179227 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179594 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32135025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2462-84-2
Molekylformel C19 H36 O2
Molekylvikt (g/mol) 296.49
Synonym 9-Octadecenoic Acid Methyl Ester, 9-Octadecenoic acid, methyl ester (7CI, 9CI, ACI), Methyl 9-octadecenoate, 9-Octadecenoic acid methyl ester
IUPAC-namn methyl octadec-9-enoate
LEDER CCCCCCCCC=CCCCCCCCC(=O)OC
Specifikationer
Kemiskt namn eller material Methyl 9-Octadecenoate
Kvantitet 25 mg
InChI formel InChI=1S/C19H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h10-11H,3-9,12-18H2,1-2H3
Formel vikt 296.2715