missing translation for 'onlineSavingsMsg'
Learn More

Methyl a-Bromo-2-chlorophenylacetate, TRC

Product Code. 30478716
Change view
Click to view available options
Kvantitet:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
30478716 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478716 Supplier Toronto Research Chemicals Supplier No. TRCM2942102.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Methyl a-Bromo-2-chlorophenylacetate
CAS 85259-19-4
Molekylformel C9 H8 Br Cl O2
Kvantitet 2.5 g
Synonym Methyl A-Bromo-2-Chlorophenylacetate
InChI formel InChI=1S/C9H8BrClO2/c1-13-9(12)8(10)6-4-2-3-5-7(6)11/h2-5,8H,1H3
LEDER COC(=O)C(Br)c1ccccc1Cl
IUPAC-namn methyl 2-bromo-2-(2-chlorophenyl)acetate
Molekylvikt (g/mol) 263.52
Formel vikt 261.9396
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.