Läs mer
Methyl alpha-Cyclopentylmandelate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Methyl (2RS)-2-Cyclopentyl-2-hydroxy-2-phenylacetate |
| CAS | 19833-96-6 |
| Molekylformel | C14 H18 O3 |
| Kvantitet | 1 g |
| Synonym | Benzeneacetic acid, α-cyclopentyl-α-hydroxy-, methyl ester, Mandelic acid, α-cyclopentyl-, methyl ester (8CI), Methyl (±)-cyclopentylmandelate, Methyl 2-Cyclopentyl-2-hydroxy-2-phenylacetate, Methyl cyclopentylphenylglycolate, Methyl α-cyclopentylmandelate, NSC 93811 |
| InChI formel | InChI=1S/C14H18O3/c1-17-13(15)14(16,12-9-5-6-10-12)11-7-3-2-4-8-11/h2-4,7-8,12,16H,5-6,9-10H2,1H3 |
| LEDER | COC(=O)C(O)(C1CCCC1)c2ccccc2 |
| IUPAC-namn | methyl 2-cyclopentyl-2-hydroxy-2-phenylacetate |
| Molekylvikt (g/mol) | 234.29 |
| Formel vikt | 234.1256 |