Läs mer
Methyl alpha-D-Glucopyranoside 2,3,4,6-tetra-O-sulfate, Potassium Salt (Technical Grade), TRC
Produktkod. 30470806
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30470806 250 mg
1 options
Produktkod. 30470806 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM311500250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl alpha-D-Glucopyranoside 2,3,4,6-tetra-O-sulfate potassium salt
CAS 359437-01-7
Molekylformel C7 H10 O18 S4 . 4 K
Kvantitet 250 mg
InChI formel InChI=1S/C7H14O18S4.4K/c1-20-7-6(25-29(17,18)19)5(24-28(14,15)16)4(23-27(11,12)13)3(22-7)2-21-26(8,9)10;;;;/h3-7H,2H2,1H3,(H,8,9,10)(H,11,12,13)(H,14,15,16)(H,17,18,19);;;;/q;4*+1/p-4/t3-,4-,5+,6-,7+;;;;/m1..../s1
LEDER [K+].[K+].[K+].[K+].CO[C@H]1O[C@H](COS(=O)(=O)[O-])[C@@H](OS(=O)(=O)[O-])[C@H](OS(=O)(=O)[O-])[C@H]1OS(=O)(=O)[O-]
IUPAC-namn tetrapotassium;[(2S,3R,4S,5R,6R)-2-methoxy-3,5-disulfonatooxy-6-(sulfonatooxymethyl)oxan-4-yl] sulfate
Molekylvikt (g/mol) 666.8
Formel vikt 665.7298