Läs mer
Methyl alpha-L-Idopyranosiduronic Acid Sodium Salt, TRC
Produktkod. 30466063
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30466063 5 mg
1 options
Produktkod. 30466063 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3129305MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl alpha-L-Idopyranosiduronic Acid Sodium Salt
CAS 134355-31-0
Molekylformel C7 H11 O7 . Na
Kvantitet 5 mg
Synonym 1-O-Methyl-α-L-iduronate Monosodium Salt, Methyl-α-L-iduronic Acid Sodium Salt
InChI formel InChI=1S/C7H12O7.Na/c1-13-7-4(10)2(8)3(9)5(14-7)6(11)12;/h2-5,7-10H,1H3,(H,11,12);/q;+1/p-1/t2-,3-,4+,5+,7+;/m0./s1
LEDER [Na+].CO[C@@H]1O[C@H]([C@@H](O)[C@H](O)[C@H]1O)C(=O)[O-]
IUPAC-namn sodium;(2R,3S,4S,5R,6R)-3,4,5-trihydroxy-6-methoxyoxane-2-carboxylate
Molekylvikt (g/mol) 230.148
Formel vikt 230.04