missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Methyl Amino(2-bromophenyl)acetate Hydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Methyl Amino(2-bromophenyl)acetate Hydrochloride |
| CAS | 1219408-44-2 |
| Molekylformel | C9H10BrNO2 . HCl |
| Kvantitet | 100 mg |
| Synonym | Benzeneacetic acid, α-amino-2-bromo-, methyl ester, hydrochloride (1:1) (ACI), Methyl α-amino-2-bromobenzeneacetate hydrochloride, α-Amino-2-bromobenzeneacetic acid methyl ester hydrochloride, Methyl Amino(2-bromophenyl)acetate hydrochloride, 2-Amino-2-(2-bromophenyl)acetic acid methyl ester hydrochloride |
| InChI formel | InChI=1S/C9H10BrNO2.ClH/c1-13-9(12)8(11)6-4-2-3-5-7(6)10;/h2-5,8H,11H2,1H3;1H |
| LEDER | Cl.COC(=O)C(N)c1ccccc1Br |
| IUPAC-namn | methyl 2-amino-2-(2-bromophenyl)acetate;hydrochloride |
| Molekylvikt (g/mol) | 280.54 |
| Formel vikt | 278.9662 |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?