Läs mer
Methyl Amino(2-bromophenyl)acetate Hydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Methyl Amino(2-bromophenyl)acetate Hydrochloride |
| CAS | 1219408-44-2 |
| Molekylformel | C9H10BrNO2 . HCl |
| Kvantitet | 1 g |
| Synonym | Benzeneacetic acid, α-amino-2-bromo-, methyl ester, hydrochloride (1:1) (ACI), Methyl α-amino-2-bromobenzeneacetate hydrochloride, α-Amino-2-bromobenzeneacetic acid methyl ester hydrochloride, Methyl Amino(2-bromophenyl)acetate hydrochloride, 2-Amino-2-(2-bromophenyl)acetic acid methyl ester hydrochloride |
| InChI formel | InChI=1S/C9H10BrNO2.ClH/c1-13-9(12)8(11)6-4-2-3-5-7(6)10;/h2-5,8H,11H2,1H3;1H |
| LEDER | Cl.COC(=O)C(N)c1ccccc1Br |
| IUPAC-namn | methyl 2-amino-2-(2-bromophenyl)acetate;hydrochloride |
| Molekylvikt (g/mol) | 280.54 |
| Formel vikt | 278.9662 |