missing translation for 'onlineSavingsMsg'
Läs mer

Methyl Atrarate, TRC

Produktkod. 30479285
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30479285 5 g
30472466 25 g
30477411 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30479285 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2883405G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 4707-47-5
Molekylformel C10 H12 O4
Molekylvikt (g/mol) 196.1999
Synonym Atraric acid (6CI), β-Resorcylic acid, 3,6-dimethyl-, methyl ester (7CI,8CI), 2,4-Dihydroxy-3,6-dimethyl-benzoic acid methyl ester, Everniate, Evernyl, LRG 201, Methyl 2,4-dihydroxy-3,6-dimethylbenzoate, Methyl 3,6-dimethyl-2,4-dihydroxybenzoate, Methyl 3,6-dimethyl-β-resorcylate, Methyl atrarate, Methyl β-orcinolcarboxylate, Methyl β-orsellinate, Moss Synth, Mousse cristal, Oakmoss synthetic, Veramoss, 2,4-Dihydroxy-3,6-dimethylbenzoic Acid Methyl Ester, Atraric acid-methyl ester, Methyl atratate
IUPAC-namn methyl 2,4-dihydroxy-3,6-dimethylbenzoate
LEDER COC(=O)c1c(C)cc(O)c(C)c1O

Specifications

Kemiskt namn eller material 2,4-Dihydroxy-3,6-dimethylbenzoic Acid Methyl Ester
Kvantitet 5 g
InChI formel InChI=1S/C10H12O4/c1-5-4-7(11)6(2)9(12)8(5)10(13)14-3/h4,11-12H,1-3H3
Formel vikt 196.0736
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.