Läs mer
Methyl b-D-arabinopyranoside, TRC
Produktkod. 30475746
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30475746 250 mg
1 options
Produktkod. 30475746 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM365155250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl-beta-D-arabinopyranoside
CAS 5328-63-2
Molekylformel C6 H12 O5
Kvantitet 250 mg
Synonym Methyl β-D-arabinopyranoside
InChI formel InChI=1S/C6H12O5/c1-10-6-5(9)4(8)3(7)2-11-6/h3-9H,2H2,1H3/t3-,4-,5+,6-/m1/s1
LEDER CO[C@@H]1OC[C@@H](O)[C@@H](O)[C@@H]1O
IUPAC-namn (2R,3S,4R,5R)-2-methoxytetrahydropyran-3,4,5-triol
Molekylvikt (g/mol) 164.16
Formel vikt 164.0685