Läs mer
Methyl Bis-anhydro-Daunomycinone Carboxylate, TRC
Produktkod. 30405705
Change view
Kvantitet:
0.5 mg
Produktkod. Kvantitet
30405705 0.5 mg
1 options
Produktkod. 30405705 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH2910680.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl Bis-anhydro-Daunomycinone Carboxylate
CAS 71809-95-5
Molekylformel C21H14O7
Kvantitet 0.5 mg
Synonym Methyl 5,12-Dihydroxy-7-methoxy-6,11-dioxo-6,11-dihydrotetracene-2-carboxylate
InChI formel InChI=1S/C21H14O7/c1-27-13-5-3-4-11-14(13)20(25)16-15(18(11)23)19(24)12-8-9(21(26)28-2)6-7-10(12)17(16)22/h3-8,22,24H,1-2H3
LEDER COC(=O)c1ccc2c(O)c3C(=O)c4c(OC)cccc4C(=O)c3c(O)c2c1
IUPAC-namn methyl 5,12-dihydroxy-7-methoxy-6,11-dioxotetracene-2-carboxylate
Molekylvikt (g/mol) 378.33
Formel vikt 378.074