Läs mer
Methyl Cyanoformate, TRC
Produktkod. 30467970
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30467970 1 g
1 options
Produktkod. 30467970 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2952501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl Cyanoformate
CAS 17640-15-2
Molekylformel C3H3NO2
Kvantitet 1 g
Synonym Cyanoformic Acid Methyl Ester, Mander's Reagent, Methyl Carbonocyanidate, Methyl Cyanoformate
InChI formel InChI=1S/C3H3NO2/c1-6-3(5)2-4/h1H3
LEDER O=C(OC)C#N
IUPAC-namn methyl carbonocyanidate
Molekylvikt (g/mol) 85.06
Formel vikt 85.02