Läs mer
Methyl Cyclohexanepropanoate, TRC
Produktkod. 30466202
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30466202 100 mg
1 options
Produktkod. 30466202 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM338915100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl Cyclohexanepropanoate
CAS 20681-51-0
Molekylformel C10 H18 O2
Kvantitet 100 mg
Synonym Methyl 3-Cyclohexylpropanoate, Methyl 3-Cyclohexylpropionate, Methyl Cyclohexanepropionate, NSC 71455
InChI formel InChI=1S/C10H18O2/c1-12-10(11)8-7-9-5-3-2-4-6-9/h9H,2-8H2,1H3
LEDER COC(=O)CCC1CCCCC1
IUPAC-namn methyl 3-cyclohexylpropanoate
Molekylvikt (g/mol) 170.249
Formel vikt 170.131