Läs mer
Methyl-D-erythritol Phosphate Benzyl Ether Dibenzyl Ester, TRC
Produktkod. 30478640
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30478640 10 mg
1 options
Produktkod. 30478640 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM30465510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl-D-erythritol Phosphate Benzyl Ether Dibenzyl Ester
CAS 332888-40-1
Molekylformel C26H31O7P
Kvantitet 10 mg
Synonym (2R,3S)-2,3-Dihydroxy-3-methyl-4-phosphoric Acid (Phenylmethoxy)butyl Bis(phenylmethyl) Ester
InChI formel InChI=1S/C26H31O7P/c1-26(28,21-30-17-22-11-5-2-6-12-22)25(27)20-33-34(29,31-18-23-13-7-3-8-14-23)32-19-24-15-9-4-10-16-24/h2-16,25,27-28H,17-21H2,1H3/t25-,26+/m1/s1
LEDER C[C@@](COCC1=CC=CC=C1)(O)[C@H](O)COP(OCC2=CC=CC=C2)(OCC3=CC=CC=C3)=O
IUPAC-namn dibenzyl ((2R,3S)-4-(benzyloxy)-2,3-dihydroxy-3-methylbutyl) phosphate
Molekylvikt (g/mol) 486.49
Formel vikt 486.18