Läs mer
Methyl-D-erythritol Phosphate Disodium Salt, TRC
Produktkod. 30471575
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30471575 5 mg
1 options
Produktkod. 30471575 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3046505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl-D-erythritol Phosphate Disodium Salt
CAS 270928-69-3
Molekylformel C5H11Na2O7P
Kvantitet 5 mg
Synonym (2S,3R)-2-Methyl-1,2,3,4-butanetetrol 4-(Dihydrogen Phosphate) Disodium Salt, 2-C-Methyl-D-erythritol 4-phosphate Disodium Salt, MEP Disodium Salt
InChI formel InChI=1S/C5H13O7P.2Na/c1-5(8,3-6)4(7)2-12-13(9,10)11;;/h4,6-8H,2-3H2,1H3,(H2,9,10,11);;/q;2*+1/p-2
LEDER [Na+].[Na+].CC(O)(CO)C(O)COP(=O)([O-])[O-]
IUPAC-namn disodium;(2,3,4-trihydroxy-3-methylbutyl) phosphate
Molekylvikt (g/mol) 260.09
Formel vikt 260.0038