missing translation for 'onlineSavingsMsg'
Learn More

Methyl-α-D-galactopyranoside, 98+%, Thermo Scientific Chemicals

Catalog Number p-3779115
Click to view available options
Quantity:
2.5 g
10 g
50 g
This item is not returnable. View return policy
3396-99-4
MFCD00064085
76935
CHEBI:55507
This item is not returnable. View return policy
3396-99-4
MFCD00064085
76935
CHEBI:55507

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 116.0°C to 117.0°C
Assay Percent Range 98% min. (ELSD) (HPLC)
Packaging Glass bottle
Molecular Formula C7H14O6
Quantity 2.5 g
Specific Rotation Condition + 179.00 (20.00°C c=1,water)
Specific Rotation + 179.00
Synonym me-alpha-gal, methyl-alpha-d-galactopyranoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-methoxytetrahydro-2h-pyran-3,4,5-triol, methyl-galactopyranoside, methyl a-d-galactopyranoside, methyl alpha-d-galactoside, alpha-methylgalactoside, alpha-methyl d-galactoside, alpha-methyl-d-galactoside, methyl alpha-d-galactopyranoside
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol
InChI Key HOVAGTYPODGVJG-UHFFFAOYNA-N
SMILES COC1OC(CO)C(O)C(O)C1O
IUPAC Name 2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
Molecular Weight (g/mol) 194.18
Formula Weight 194.18
Percent Purity 98+%
Chemical Name or Material Methyl-α-D-galactopyranoside
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'