Läs mer
Methyl Dehydroabietate, TRC
Produktkod. 30136853
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136853 1 g
30007230 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136853 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2719201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1235-74-1
Molekylformel C18 H24 O2
Molekylvikt (g/mol) 272.38
Synonym (1R,4aS,10aR)-1,2,3,4,4a,9,10,10a-Octahydro-1,4a-dimethyl-7-(1-methylethyl)-1-phenanthrenecarboxylic Acid Methyl Ester;
IUPAC-namn methyl (1R,4aS,10aR)-1,4a-dimethyl-2,3,4,9,10,10a-hexahydrophenanthrene-1-carboxylate
LEDER COC(=O)[C@]1(C)CCC[C@@]2(C)[C@H]1CCc3ccccc23
Specifikationer
Kemiskt namn eller material Methyl Dehydroabietate
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C18H24O2/c1-17-11-6-12-18(2,16(19)20-3)15(17)10-9-13-7-4-5-8-14(13)17/h4-5,7-8,15H,6,9-12H2,1-3H3/t15-,17-,18-/m1/s1
Formel vikt 272.1776
Procent renhet >95%