Learn More
Methyl diethylphosphonoacetate, 97%, Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
Methyl diethylphosphonoacetate is used as a reagent in the preparation of allylic fluorides compounds and in regioselective Diels-Alder reactions. It also serves as a reactant in the preparation of substituted thiophenes and furans, which finds application in type 2 diabetes treatment. Further, it acts as a precursor in the synthesis of pyridone alkaloids and immunosuppressive cyclopentanediol derivatives. In addition to this, it is utilized in the modification of botulinum neurotoxin serotype A protease inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyl diethylphosphonoacetate is used as a reagent in the preparation of allylic fluorides compounds and in regioselective Diels-Alder reactions. It also serves as a reactant in the preparation of substituted thiophenes and furans, which finds application in type 2 diabetes treatment. Further, it acts as a precursor in the synthesis of pyridone alkaloids and immunosuppressive cyclopentanediol derivatives. In addition to this, it is utilized in the modification of botulinum neurotoxin serotype A protease inhibitors.
Solubility
Miscible with tetrahydrofuran.
Notes
Incompatible with strong oxidizing agents.
Specifications
Density | 1.145 |
Boiling Point | 126°C to 128°C (8 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.432 |
Quantity | 25 g |
Beilstein | 1782397 |
Solubility Information | Miscible with tetrahydrofuran. |
Formula Weight | 210.17 |
Percent Purity | 97% |
Chemical Name or Material | Methyl diethylphosphonoacetate |
RUO – Research Use Only