Läs mer
Methyl Dl-2-(1-Boc-Piperidin-4-Yl)-Beta-Glycinate, TRC
Produktkod. 30471433
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30471433 100 mg
30474263 10 mg
30475903 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30471433 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM315505100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 362703-57-9
Molekylformel C13H24N2O4
Molekylvikt (g/mol) 272.34
Synonym Methyl 4-Amino-1-[(1,1-dimethylethoxy)carbonyl]-4-piperidineacetate, tert-Butyl 4-amino-4-(2-methoxy-2-oxoethyl)piperidine-1-carboxylate
IUPAC-namn tert-butyl 4-amino-4-(2-methoxy-2-oxoethyl)piperidine-1-carboxylate
LEDER COC(=O)CC1(N)CCN(CC1)C(=O)OC(C)(C)C
Specifikationer
Kemiskt namn eller material Methyl Dl-2-(1-Boc-Piperidin-4-Yl)-beta-Glycinate
Kvantitet 100 mg
InChI formel InChI=1S/C13H24N2O4/c1-12(2,3)19-11(17)15-7-5-13(14,6-8-15)9-10(16)18-4/h5-9,14H2,1-4H3
Formel vikt 272.1736