Läs mer
Methyl Ergonovine Maleate Salt, TRC
Produktkod. 30008071
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008071 5 mg
30005265 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30008071 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3046005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 57432-61-8
Molekylformel C20 H25 N3 O2 . C4 H4 O4
Molekylvikt (g/mol) 455.5
Synonym Methylergometrine maleate, (6aR,9R)-N-[(1S)-1-(Hydroxymethyl)propyl]-7-methyl- 4,6,6a,7,8,9-hexahydroindolo[4,3-fg]quinoline-9-carboxamide (Z)-butenedioate
IUPAC-namn (6aR,9R)-N-[(2S)-1-hydroxybutan-2-yl]-7-methyl-6,6a,8,9-tetrahydro-4H-indolo[4,3-fg]quinoline-9-carboxamide;(Z)-but-2-enedioic acid
LEDER CC[C@@H](CO)NC(=O)[C@H]1CN(C)[C@@H]2Cc3c[nH]c4cccc(C2=C1)c34.OC(=O)\C=C/C(=O)O
Specifikationer
Kemiskt namn eller material Methylergometrine maleate
Kvantitet 5 mg
InChI formel InChI=1S/C20H25N3O2.C4H4O4/c1-3-14(11-24)22-20(25)13-7-16-15-5-4-6-17-19(15)12(9-21-17)8-18(16)23(2)10-13;5-3(6)1-2-4(7)8/h4-7,9,13-14,18,21,24H,3,8,10-11H2,1-2H3,(H,22,25);1-2H,(H,5,6)(H,7,8)/b;2-1-/t13-,14+,18-;/m1./s1
Formel vikt 455.2056