Läs mer
Methyl Formate-d, TRC
Produktkod. 30471777
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30471777 1 g
1 options
Produktkod. 30471777 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3053921G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl Formate-d
CAS 23731-38-6
Molekylformel C2 D H3 O2
Kvantitet 1 g
Synonym Formic-d acid','methyl ester (6CI','7CI','8CI','9CI), Methyl deuterioformate, Methyl Formate-d1, Methyl Formate-D, Formic-d acid methyl ester, Formic acid methyl ester-d1
InChI formel InChI=1S/C2H4O2/c1-4-2-3/h2H,1H3/i2D
LEDER [2H]C(=O)OC
IUPAC-namn methyl deuterioformate
Molekylvikt (g/mol) 61.0581
Formel vikt 61.0274