Läs mer
Methyl Heptadecanoate, TRC
Produktkod. 30472970
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30472970 1 g
1 options
Produktkod. 30472970 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3118851G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Heptadecanoic acid-methyl ester
CAS 1731-92-6
Molekylformel C18 H36 O2
Kvantitet 1 g
Synonym Heptadecanoic acid-methyl ester, Heptadecanoic Acid Methyl Ester, Heptadecanoic acid','methyl ester (6CI','7CI','8CI','9CI','ACI), A 27667, Margaric acid methyl ester, Methyl heptadecanoate, Methyl margarate, NSC 97364
InChI formel InChI=1S/C18H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20-2/h3-17H2,1-2H3
LEDER CCCCCCCCCCCCCCCCC(=O)OC
IUPAC-namn methyl heptadecanoate
Molekylvikt (g/mol) 284.48
Formel vikt 284.2715