Läs mer
Methyl hydrogen azelate, 96%
Produktkod. p-7044761
Kvantitet:
5 g
25 g
Förpackningsstorlek:
25g
5g
Produktkod. 11307995
Brand: Thermo Scientific Alfa Aesar L08116.06
Request Bulk or Custom Format
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
CAS: 2104-19-0 | C10H18O4 | 202.25 g/mol
Methyl hydrogen azelate is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl hydrogen azelate is used as pharmaceutical intermediates.
Solubility
Not miscible in water.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2104-19-0
Molekylformel C10H18O4
Molekylvikt (g/mol) 202.25
MDL-nummer MFCD00004431
InChI-nyckel VVWPSAPZUZXYCM-UHFFFAOYSA-N
Synonym methyl hydrogen azelate, monomethyl azelate, azelaic acid monomethyl ester, nonanedioic acid, monomethyl ester, monomethyl nonanedioate, 8-carbomethoxyoctanoic acid, azelaic acid, monomethyl ester, mono-methyl azelate, unii-xdi67hf5bh, xdi67hf5bh
PubChem CID 75009
IUPAC-namn 9-methoxy-9-oxononanoic acid
LEDER COC(=O)CCCCCCCC(=O)O
Specifikationer
Kemiskt namn eller material Methyl hydrogen azelate
Smältpunkt 22°C to 24°C
Kokpunkt 146°C to 147°C (1 mmHg)
Flampunkt >110°C (230°F)
Linjär formel CH3OOC(CH2)7COOH
Brytningsindex 1.447
Kvantitet 5 g
Beilstein 1775786
Löslighetsinformation Not miscible in water.
Formel vikt 202.25
Procent renhet 96%