missing translation for 'onlineSavingsMsg'
Learn More

Methyl isocyanoacetate, 95%, Tech., Thermo Scientific Chemicals

Catalog Number p-3736925
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
39687-95-1
C4H7NO2
101.11
MFCD00000006
ZRGVTTCCASYOFN-UHFFFAOYSA-N
methyl isocyanoacetate, isocyanoacetic acid methyl ester, methylisocyanoacetate, isocyano-acetic acid methyl ester, acetic acid,2-isocyano-, methyl ester, methyl-isocyanoacetate, methyl isocyano-acetate, acmc-20akf8, methyl alpha-isocyanoacetate, ksc581o3d
547815
methyl 2-isocyanoacetate
COC(=O)CN=C
This item is not returnable. View return policy
39687-95-1
C4H7NO2
101.11
MFCD00000006
ZRGVTTCCASYOFN-UHFFFAOYSA-N
methyl isocyanoacetate, isocyanoacetic acid methyl ester, methylisocyanoacetate, isocyano-acetic acid methyl ester, acetic acid,2-isocyano-, methyl ester, methyl-isocyanoacetate, methyl isocyano-acetate, acmc-20akf8, methyl alpha-isocyanoacetate, ksc581o3d
547815
methyl 2-isocyanoacetate
COC(=O)CN=C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow to Brown
Density 1.0900g/mL
Boiling Point 75°C to 76°C (10.0 mmHg)
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula CNCH2CO2CH3
Refractive Index 1.4161 to 1.4191
Quantity 5 g
Specific Gravity 1.09
Formula Weight 99.09
Percent Purity 95%
Grade Technical
Physical Form Liquid
Chemical Name or Material Methyl isocyanoacetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'