Läs mer
Methyl L-Daunosamine Hydrochloride, TRC
Produktkod. 30479175
Change view
Kvantitet:
2.5 mg
Produktkod. Kvantitet
30479175 2.5 mg
1 options
Produktkod. 30479175 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2948052.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl L-Daunosamine Hydrochloride
CAS 131528-45-5
Molekylformel C7H16ClNO3
Kvantitet 2.5 mg
Synonym Methyl 3-Amino-2,3,6-trideoxy-L-lyxo-hexopyranoside Hydrochloride
InChI formel InChI=1S/C7H15NO3.ClH/c1-4-7(9)5(8)3-6(10-2)11-4;/h4-7,9H,3,8H2,1-2H3;1H/t4-,5-,6?,7+;/m0./s1
LEDER COC1C[C@@H]([C@@H]([C@@H](O1)C)O)N.Cl
IUPAC-namn (2S,3S,4S)-4-amino-6-methoxy-2-methyltetrahydro-2H-pyran-3-ol hydrochloride
Molekylvikt (g/mol) 197.66
Formel vikt 197.08