Läs mer
Methyl N-Acetylglycinate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Methyl N-Acetylglycinate |
| CAS | 1117-77-7 |
| Molekylformel | C5 H9 N O3 |
| Kvantitet | 5 g |
| Synonym | Glycine','N-acetyl-','methyl ester, Aceturic acid','methyl ester (6CI), Methyl 2-acetamidoacetate, Methyl N-acetylglycinate, Methyl N-acetylglycine, Methyl acetylglycinate, N-Acetylglycine methyl ester |
| InChI formel | InChI=1S/C5H9NO3/c1-4(7)6-3-5(8)9-2/h3H2,1-2H3,(H,6,7) |
| LEDER | COC(=O)CNC(=O)C |
| IUPAC-namn | methyl 2-acetamidoacetate |
| Molekylvikt (g/mol) | 131.13 |
| Formel vikt | 131.0582 |