Läs mer
Methyl N-Methoxy-2-methylphenylcarbamate, TRC
Produktkod. 30475362
Change view
Kvantitet:
2.5 g
Produktkod. Kvantitet
30475362 2.5 g
1 options
Produktkod. 30475362 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2587252.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl N-Methoxy-2-methylphenylcarbamate
CAS 151827-82-6
Molekylformel C10H13NO3
Kvantitet 2.5 g
Synonym Methyl N-Methoxy-N-2-methylphenylcarbamate
InChI formel InChI=1S/C10H13NO3/c1-8-6-4-5-7-9(8)11(14-3)10(12)13-2/h4-7H,1-3H3
LEDER CON(C(=O)OC)c1ccccc1C
Molekylvikt (g/mol) 195.22
Formel vikt 195.0895