Läs mer
Methyl (R)-2-[(tert-Butyldimethylsilyl)oxy]-2-(3-chlorophenyl)acetate, TRC
Produktkod. 30478855
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30478855 1 g
1 options
Produktkod. 30478855 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3051881G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl (R)-2-[(tert-Butyldimethylsilyl)oxy]-2-(3-chlorophenyl)acetate
CAS 153294-01-0
Molekylformel C15H23ClO3Si
Kvantitet 1 g
Synonym (alphaR)-3-Chloro-alpha-[[(1,1-dimethylethyl)dimethylsilyl]oxy]-benzeneacetic Acid Methyl Ester, (R)- 3-Chloro-alpha-[[(1,1-dimethylethyl)dimethylsilyl]oxy]-benzeneacetic Acid Methyl Ester
InChI formel InChI=1S/C15H23ClO3Si/c1-15(2,3)20(5,6)19-13(14(17)18-4)11-8-7-9-12(16)10-11/h7-10,13H,1-6H3/t13-/m1/s1
LEDER COC(=O)[C@H](O[Si](C)(C)C(C)(C)C)c1cccc(Cl)c1
IUPAC-namn methyl (2R)-2-[tert-butyl(dimethyl)silyl]oxy-2-(3-chlorophenyl)acetate
Molekylvikt (g/mol) 314.88
Formel vikt 314.1105