missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Red, Pure, Spirit Soluble, C.I.13020, Indicator Grade, Fisher Chemical™
Click to view available options
Quantity:
10 g
25 g
Specifications
Melting Point | 178°C |
Formula Weight | 269.3g/mol |
Packaging | Glass Jar |
Color | Red |
Quantity | 10 g |
Physical Form | Solid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'