Läs mer
Methyl (RS)-(2,2,2-Trichloro-1-hydroxyethyl)phosphonate Acid (Desmethylmetrifonate), Mikromol™
Beskrivning
Methyl (RS)-(2,2,2-Trichloro-1-hydroxyethyl)phosphonate Acid (Desmethylmetrifonate)
Specifikationer
Specifikationer
| Kemiskt namn eller material | Desmethylmetrifonate |
| CAS | 684-31-1 |
| Analyt- eller komponentnamn | Desmethylmetrifonate |
| Frakt skick | Room Temperature |
| Molekylformel | C3 H6 Cl3 O4 P |
| Certifikat och efterlevnad | ISO 9001 |
| Synonym | Phosphonic acid, P-(2,2,2-trichloro-1-hydroxyethyl)-, monomethyl ester, Phosphonic acid, (2,2,2-trichloro-1-hydroxyethyl)-, methyl ester (7CI), Phosphonic acid, (2,2,2-trichloro-1-hydroxyethyl)-, monomethyl ester (8CI,9CI), Demethyltrichlorfon, Demethyltrichlorphon, Desmethyltrichlorphone, Monomethyltrichlorphon, O-Methyl-2,2,2-trichloro-1-hydroxyethylphosphonate, Methyl (RS)-(2,2,2-Trichloro-1-hydroxyethyl)phosphonate Acid |
| InChI formel | InChI=1S/C3H6Cl3O4P/c1-10-11(8,9)2(7)3(4,5)6/h2,7H,1H3,(H,8,9) |
| LEDER | COP(=O)(O)C(O)C(Cl)(Cl)Cl |
| IUPAC-namn | methoxy-(2,2,2-trichloro-1-hydroxyethyl)phosphinic acid |
| Visa mer |