Läs mer
Methyl [(RS)-(Diphenylmethyl)sulfinyl]acetate, Mikromol™
Produktkod. 30051976
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30051976 50 mg
1 options
Produktkod. 30051976 Leverantör Mikromol Leverantörsnummer MM0540.03
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Methyl [(RS)-(Diphenylmethyl)sulfinyl]acetate
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl [(RS)-(Diphenylmethyl)sulfinyl]acetate
CAS 63547-25-1
Analyt- eller komponentnamn Methyl [(RS)-(Diphenylmethyl)sulfinyl]acetate
Frakt skick Room Temperature
Molekylformel C16 H16 O3 S
Certifikat och efterlevnad ISO 17025
Synonym Methyl [(RS)-(Diphenylmethyl)sulfinyl]acetate, Modafinil Imp. C (EP), Modafinil Impurity C
InChI formel InChI=1S/C16H16O3S/c1-19-15(17)12-20(18)16(13-8-4-2-5-9-13)14-10-6-3-7-11-14/h2-11,16H,12H2,1H3
LEDER COC(=O)CS(=O)C(c1ccccc1)c2ccccc2
IUPAC-namn methyl 2-benzhydrylsulfinylacetate
Molekylvikt (g/mol) 288.36
Kvantitet 50 mg
Formel vikt 288.082 g/mol