Läs mer
Methyl ((S)-4-Methyl-2-[2-oxo-2-[[(S)-1-phenylethyl]amino]ethyl]pentyl]carbamate, TRC
Produktkod. 30484016
Change view
Kvantitet:
5 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30484016 100 mg
30467361 250 mg
30469134 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30484016 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM322015100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 930280-44-7
Molekylformel C18H28N2O3
Molekylvikt (g/mol) 320.43
Synonym N-[(2S)-4-Methyl-2-[2-oxo-2-[[(1S)-1-phenylethyl]amino]ethyl]pentyl]carbamic Acid Methyl Ester, Carbamic acid, N-[(2S)-4-methyl-2-[2-oxo-2-[[(1S)-1-phenylethyl]amino]ethyl]pentyl]-, methyl ester, Methyl N-[(2S)-4-methyl-2-[2-oxo-2-[[(1S)-1-phenylethyl]amino]ethyl]pentyl]carbamate, N-[(2S)-4-Methyl-2-[2-oxo-2-[[(1S)-1-phenylethyl]amino]ethyl]pentyl]carbamic Acid Methyl Ester, Carbamic acid, N-[(2S)-4-methyl-2-[2-oxo-2-[[(1S)-1-phenylethyl]amino]ethyl]pentyl]-, methyl ester, Methyl N-[(2S)-4-methyl-2-[2-oxo-2-[[(1S)-1-phenylethyl]amino]ethyl]pentyl]carbamate
IUPAC-namn methyl N-[(2S)-4-methyl-2-[2-oxo-2-[[(1S)-1-phenylethyl]amino]ethyl]pentyl]carbamate
LEDER COC(=O)NC[C@@H](CC(C)C)CC(=O)N[C@@H](C)c1ccccc1
Specifikationer
Kemiskt namn eller material Methyl ((S)-4-Methyl-2-(2-oxo-2-(((S)-1-phenylethyl)amino)ethyl)pentyl)carbamate
Kvantitet 100 mg
InChI formel InChI=1S/C18H28N2O3/c1-13(2)10-15(12-19-18(22)23-4)11-17(21)20-14(3)16-8-6-5-7-9-16/h5-9,13-15H,10-12H2,1-4H3,(H,19,22)(H,20,21)/t14-,15-/m0/s1
Formel vikt 320.21