Läs mer
Metylsalicylat beta-D-O-glucuronidtriacetat metylester, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Methyl Salicylate beta-D-O-Glucuronide Triacetate Methyl Ester |
| CAS | 101231-54-3 |
| Molekylformel | C21H24O12 |
| Kvantitet | 25 mg |
| Synonym | 2-(Methoxycarbonyl)phenyl beta-D-Glucopyranosiduronic Acid Methyl Triacetate Ester, O-carboxyphenyl beta-D-Glucopyranosiduronic alphacid Triacetate Dimethyl Ester. |
| InChI formel | InChI=1S/C21H24O12/c1-10(22)29-15-16(30-11(2)23)18(31-12(3)24)21(33-17(15)20(26)28-5)32-14-9-7-6-8-13(14)19(25)27-4/h6-9,15-18,21H,1-5H3/t15-,16-,17-,18+,21+/m0/s1 |
| LEDER | COC(=O)[C@H]1O[C@@H](Oc2ccccc2C(=O)OC)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C |
| IUPAC-namn | metyl (2S,3S,4S,5R,6S)-3,4,5-triacetyloxi-6-(2-metoxykarbonylfenoxi)oxan-2-karboxyllat |
| Molekylvikt (g/mol) | 468.41 |
| Formel vikt | 468.1268 |