Läs mer
Methyl trans-10-Octadecenoate, TRC
Produktkod. 30138345
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138345 10 mg
30005491 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138345 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM27304010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13038-45-4
Molekylformel C19 H36 O2
Molekylvikt (g/mol) 296.488
IUPAC-namn methyl (E)-octadec-10-enoate
LEDER CCCCCCC\C=C\CCCCCCCCC(=O)OC
Specifikationer
Kemiskt namn eller material Methyl trans-10-Octadecenoate
Kvantitet 10 mg
InChI formel InChI=1S/C19H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h9-10H,3-8,11-18H2,1-2H3/b10-9+
Formel vikt 296.272