missing translation for 'onlineSavingsMsg'
Läs mer

Methyl)triphenylphosphonium Iodide-13C, TRC

Produktkod. 30468686
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30468686 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30468686 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM330999250MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material (Methyl)triphenylphosphonium Iodide-13C
CAS 81826-67-7
Molekylformel C19H18IP
Kvantitet 250 mg
Synonym ([13C]-Methyl)triphenylphosphonium Iodide
InChI formel InChI=1S/C19H18P.HI/c1-20(17-11-5-2-6-12-17,18-13-7-3-8-14-18)19-15-9-4-10-16-19;/h2-16H,1H3;1H/q+1;/p-1/i1+1;
LEDER [I-].[13CH3][P+](c1ccccc1)(c2ccccc2)c3ccccc3
IUPAC-namn (1^{13}C)methyl(triphenyl)phosphanium;iodide
Molekylvikt (g/mol) 405.2
Formel vikt 405.0224
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.