Learn More
Dimethyl trisulfide, 98+%, Thermo Scientific Chemicals
Catalog Number p-169351
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3658-80-8
C2H6S3
126.27
MFCD00039808
YWHLKYXPLRWGSE-UHFFFAOYSA-N
dimethyl trisulfide, trisulfide, dimethyl, methyl trisulfide, 2,3,4-trithiapentane, dimethyl trisulphide, dimethyltrisulfide, methyltrisulfanyl methane, dimethyltrisulfane, unii-3e691t3nl1, dmts
19310
CHEBI:4614
(methyltrisulfanyl)methane
CSSSC
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3658-80-8
C2H6S3
126.27
MFCD00039808
YWHLKYXPLRWGSE-UHFFFAOYSA-N
dimethyl trisulfide, trisulfide, dimethyl, methyl trisulfide, 2,3,4-trithiapentane, dimethyl trisulphide, dimethyltrisulfide, methyltrisulfanyl methane, dimethyltrisulfane, unii-3e691t3nl1, dmts
19310
CHEBI:4614
(methyltrisulfanyl)methane
CSSSC
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point -68.0°C
Color Yellow
Density 1.1900g/mL
Boiling Point 165.0°C to 175.0°C
Flash Point 52°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3SSSCH3
Refractive Index 1.5995 to 1.6025
Quantity 5 g
Beilstein 01, IV, 1283
Specific Gravity 1.19
Formula Weight 126.27
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Dimethyl trisulfide, 98+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dimethyl trisulfide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.