Läs mer
Methyl Urocanate, TRC
Produktkod. 30481523
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30481523 10 g
1 options
Produktkod. 30481523 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM33847510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl Urocanate
CAS 52363-40-3
Molekylformel C7 H8 N2 O2
Kvantitet 10 g
Synonym 2-Propenoic acid, 3-(1H-imidazol-5-yl)-, methyl ester, 2-Propenoic acid, 3-(1H-imidazol-4-yl)-, methyl ester (9CI), Imidazole-4(or 5)-acrylic acid, methyl ester (7CI), 3-(1H-Imidazol-4-yl)acrylic acid methyl ester, Methyl 3-(1H-imidazol-4-yl)acrylate, Methyl urocanate, Urocanic acid methyl ester
InChI formel InChI=1S/C7H8N2O2/c1-11-7(10)3-2-6-4-8-5-9-6/h2-5H,1H3,(H,8,9)/b3-2+
LEDER COC(=O)\C=C\c1c[nH]cn1
IUPAC-namn methyl (E)-3-(1H-imidazol-4-yl)prop-2-enoate
Molekylvikt (g/mol) 152.15
Formel vikt 152.0586