missing translation for 'onlineSavingsMsg'
Learn More

(Methylamino)acetaldehyde dimethyl acetal, 97%, Thermo Scientific Chemicals

Catalog Number p-7043731
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
122-07-6
C5H13NO2
119.164
MFCD00008485
HUMIEJNVCICTPJ-UHFFFAOYSA-N
methylaminoacetaldehyde dimethyl acetal, methylamino acetaldehyde dimethyl acetal, ethanamine, 2,2-dimethoxy-n-methyl, 2,2-dimethoxyethyl methyl amine, n-methylaminoacetaldehyde dimethyl acetal, n-2,2-dimethoxyethyl methylamine, unii-6v8q54khcl, 2,2-dimethoxyethylmethylamine, 2-methylamino acetaldehyde dimethyl acetal
8503
2,2-dimethoxy-N-methylethanamine
CNCC(OC)OC
This item is not returnable. View return policy
122-07-6
C5H13NO2
119.164
MFCD00008485
HUMIEJNVCICTPJ-UHFFFAOYSA-N
methylaminoacetaldehyde dimethyl acetal, methylamino acetaldehyde dimethyl acetal, ethanamine, 2,2-dimethoxy-n-methyl, 2,2-dimethoxyethyl methyl amine, n-methylaminoacetaldehyde dimethyl acetal, n-2,2-dimethoxyethyl methylamine, unii-6v8q54khcl, 2,2-dimethoxyethylmethylamine, 2-methylamino acetaldehyde dimethyl acetal
8503
2,2-dimethoxy-N-methylethanamine
CNCC(OC)OC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -73°C
Density 0.928
pH 10.9
Boiling Point 139°C to 140°C
Flash Point 29°C (84°F)
Odor Ammonia-like
Refractive Index 1.4115
Quantity 25 g
UN Number UN1993
Beilstein 605322
Sensitivity Air Sensitive
Formula Weight 119.16
Percent Purity 97%
Chemical Name or Material (Methylamino)acetaldehyde dimethyl acetal
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'