Läs mer
Methylbenzeneethanol, TRC
Produktkod. 30466690
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30466690 1 g
1 options
Produktkod. 30466690 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2888701G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Phenyl-2-propanol
CAS 698-87-3
Molekylformel C9 H12 O
Kvantitet 1 g
Synonym alpha-Methylbenzeneethanol, alpha-Methylphenethyl alcohol, (+/-)-1-Phenylpropan-2-ol, (+/-)-alpha-Methylbenzeneethanol, (+/-)-alpha-Methylphenethyl alcohol, 1-Phenyl-2-propanol, 2-Hydroxy-1-phenylpropane, 1-Phenyl-2-propanol, 3-Phenyl-2-hydroxypropane, 3-Phenyl-2-propanol, Benzyl methyl carbinol, Benzylethyl alcohol, NSC 53553, alpha-Methylbenzeneethanol, alpha-Methylphenethyl alcohol
InChI formel InChI=1S/C9H12O/c1-8(10)7-9-5-3-2-4-6-9/h2-6,8,10H,7H2,1H3
LEDER CC(O)Cc1ccccc1
IUPAC-namn 1-phenylpropan-2-ol
Molekylvikt (g/mol) 136.19
Formel vikt 136.0888