missing translation for 'onlineSavingsMsg'
Learn More

Methylboronic Acid-d3, TRC

Product Code. 30472572
Change view
Click to view available options
Kvantitet:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30472572 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30472572 Supplier Toronto Research Chemicals Supplier No. TRCM26509250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Methylboronic Acid-d3
CAS 1332481-37-4
Molekylformel CH2D3BO2
Kvantitet 50 mg
Synonym Methaneboronic Acid-d3, B-(Methyl-d3)boronoic Acid
InChI formel InChI=1S/CH5BO2/c1-2(3)4/h3-4H,1H3/i1D3
LEDER [2H]C([2H])([2H])B(O)O
IUPAC-namn (methyl-d3)boronic acid
Molekylvikt (g/mol) 62.88
Formel vikt 63.06
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.