Läs mer
Methyldesphenylchloridazon, TRC
Produktkod. 30011796
Change view
Kvantitet:
250 mg
1 g
2.5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30011796 2.5 g 2.5g
30005961 250 mg -
30003515 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011796 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2957802.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 17254-80-7
Molekylformel C5 H6 Cl N3 O
Molekylvikt (g/mol) 159.57
Synonym 5-Amino-4-chloro-2-methyl-3(2H)-pyridazinone, Methyldesphenylchloridazon
IUPAC-namn 5-amino-4-chloro-2-methylpyridazin-3-one
LEDER CN1N=CC(=C(Cl)C1=O)N
Specifikationer
Kemiskt namn eller material Chloridazon methyl desphenyl
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 g
InChI formel InChI=1S/C5H6ClN3O/c1-9-5(10)4(6)3(7)2-8-9/h2H,7H2,1H3
Formel vikt 159.0199
Procent renhet >95%