Läs mer
Methyldiphenylmethane, TRC
Produktkod. 30479370
Change view
Kvantitet:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30479370 250 mg
30475800 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30479370 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM312573250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 612-00-0
Molekylformel C14H14
Molekylvikt (g/mol) 182.26
Synonym 1,1'-Ethylidenebis[benzene], 1,1-Diphenylethane, NSC 33534, Pabsol 300
IUPAC-namn 1-phenylethylbenzene
LEDER CC(c1ccccc1)c2ccccc2
Specifikationer
Kemiskt namn eller material Methyldiphenylmethane
Kvantitet 250 mg
InChI formel InChI=1S/C14H14/c1-12(13-8-4-2-5-9-13)14-10-6-3-7-11-14/h2-12H,1H3
Formel vikt 182.1096