missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(hydroxymethyl)acrylate, 85%, stabilized with 0.1 wt.% MEHQ, Thermo Scientific Chemicals

Catalog Number p-4323667
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
15484-46-5, 150-19-6
C5H8O3
116.12
MFCD12545894
RFUCOAQWQVDBEU-UHFFFAOYSA-N
methyl 2-hydroxymethyl acrylate, methyl 2-hydroxymethyl prop-2-enoate, methyl2-hydroxymethyl acrylate, methyl-2-hydroxymethyl-acrylate, pubchem17373, methyl 2-hydroxymethylacrylate, methyl alpha-hydroxymethylacrylate, 2-hydroxymethylacrylate methyl ester, 2-methylolacrylic acid methyl ester, methyl alpha-hydroxymethyl acrylate
11815329
methyl 2-(hydroxymethyl)prop-2-enoate
COC(=O)C(=C)CO
This item is not returnable. View return policy
15484-46-5, 150-19-6
C5H8O3
116.12
MFCD12545894
RFUCOAQWQVDBEU-UHFFFAOYSA-N
methyl 2-hydroxymethyl acrylate, methyl 2-hydroxymethyl prop-2-enoate, methyl2-hydroxymethyl acrylate, methyl-2-hydroxymethyl-acrylate, pubchem17373, methyl 2-hydroxymethylacrylate, methyl alpha-hydroxymethylacrylate, 2-hydroxymethylacrylate methyl ester, 2-methylolacrylic acid methyl ester, methyl alpha-hydroxymethyl acrylate
11815329
methyl 2-(hydroxymethyl)prop-2-enoate
COC(=O)C(=C)CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Density 1.1290g/mL
Boiling Point 71°C to 72°C (2.0 Torr)
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 84% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 1.129
Stabilizer 0.1 wt% Monomethylether hydroquinone
Formula Weight 116.12
Percent Purity 85%
Physical Form Powder
Chemical Name or Material Methyl 2-(hydroxymethyl)acrylate, Stabilized
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 2-(hydroxymethyl)acrylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'