missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-methoxysalicylate, 98%, Thermo Scientific™

Catalog Number p-3758432
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5446-02-6
C9H10O4
182.18
MFCD00008424
ZICRWXFGZCVTBZ-UHFFFAOYSA-N
Methyle2-hydroxy-4-methoxybenzoate
methyl 2-hydroxy-4-methoxybenzoate
COC(=O)C1=CC=C(OC)C=C1O
This item is not returnable. View return policy
5446-02-6
C9H10O4
182.18
MFCD00008424
ZICRWXFGZCVTBZ-UHFFFAOYSA-N
Methyle2-hydroxy-4-methoxybenzoate
methyl 2-hydroxy-4-methoxybenzoate
COC(=O)C1=CC=C(OC)C=C1O
Melting Point 49°C to 53°C
Color White to Yellow
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H3(OH)CO2CH3
Quantity 10 g
Beilstein 10, 381
Formula Weight 182.18
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Methyl 4-methoxysalicylate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'