Learn More
Methyl cellulose, viscosity 400 cP (2% solution in water), Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Packaging | Glass bottle |
Quantity | 50 g |
Merck Index | 15,6111 |
Solubility Information | Solubility in water: soluble in cold water. Other solubilities: soluble in glacial acetic acid,insoluble in acetone,alcohol,ether,chloroform,and toluene |
Chemical Name or Material | Methyl cellulose, Viscosity 400 cP (2% solution in water) |