Läs mer
(-)-Methylephedrine Hydrochloride, TRC
Produktkod. 17923935
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17923935 1 mg
17971895 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17923935 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM8505001MG
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 38455-90-2
Molekylformel C11H17NOClH
Molekylvikt (g/mol) 215.7
Synonym (αR)-α-[(1S)-1-(Dimethylamino)ethyl]benzenemethanol hydrochloride (1:1), [R-(R*,S*)]-α-[1-(Dimethylamino)ethyl]benzenemethanol hydrochloride, (-)-Methylephedrine hydrochloride, L-erythro-Methylephedrine hydrochloride, Methy F, N-Methylephedrine hydrochloride, NSC 42289, L-N-Methylephedrine hydrochloride
IUPAC-namn (1R,2S)-2-(dimethylamino)-1-phenylpropan-1-ol;hydrochloride
LEDER Cl.C[C@@H]([C@H](O)c1ccccc1)N(C)C
Specifikationer
Kemiskt namn eller material (-)-Methylephedrine hydrochloride
Kvantitet 1 mg
InChI formel InChI=1S/C11H17NO.ClH/c1-9(12(2)3)11(13)10-7-5-4-6-8-10;/h4-9,11,13H,1-3H3;1H/t9-,11-;/m0./s1
Formel vikt 215.7197