Läs mer
Methyleugenol, Mikromol™
Produktkod. 17933526
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
17933526 100 mg
1 options
Produktkod. 17933526 Leverantör Mikromol Leverantörsnummer MM0877.19
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Methyleugenol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Eugenol Methyl Ether
CAS 93-15-2
Analyt- eller komponentnamn Eugenol Methyl Ether
Frakt skick Room Temperature
Molekylformel C11 H14 O2
Certifikat och efterlevnad ISO 17025
Synonym Benzene, 1,2-dimethoxy-4-(2-propenyl)- (9CI), Benzene, 4-allyl-1,2-dimethoxy- (8CI), Veratrole, 4-allyl- (6CI), 1,2-Dimethoxy-4-(2-propenyl)benzene, 1,2-Dimethoxy-4-allylbenzene, 1,3,4-Eugenol methyl ether, 1-(3,4-Dimethoxyphenyl)-2-propene, 1-Allyl-3,4-dimethoxybenzene, 3,4-Dimethoxy-1-(2-propenyl)benzene, 3,4-Dimethoxyallylbenzene, 3-(3,4-Dimethoxyphenyl)-1-propene, 3-(3,4-Dimethoxyphenyl)propene, 4-Allyl-1,2-dimethoxybenzene, 4-Allylveratrole, Chavibetol methyl ether, Ent 21040, Eugenol methyl ether, Eugenyl methyl ether, Methyl eugenol ether, Methyl eugenyl ether, Methylchavibetol, Methyleugenol, NSC 209528, NSC 8900, O-Methyleugenol, Veratrole methyl ether
InChI formel InChI=1S/C11H14O2/c1-4-5-9-6-7-10(12-2)11(8-9)13-3/h4,6-8H,1,5H2,2-3H3
LEDER COc1ccc(CC=C)cc1OC
IUPAC-namn 1,2-dimethoxy-4-prop-2-enylbenzene
Molekylvikt (g/mol) 178.23
Kvantitet 100 mg
Formel vikt 178.0994 g/mol