missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylglyoxal, MP Biomedicals™
Click to view available options
Quantity:
25 mL
100 mL
250 mL
Specifications
Color | Colorless |
Density | 1.17g/mL |
Quantity | 25 mL |
Formula Weight | 72.06 |
Physical Form | Liquid |
Chemical Name or Material | Methylglyoxal, Molecular biology |
Safety and Handling
Product Identifier
- Methylglyoxal, 40% Aqueous Solution
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Germ cell mutagenicity Category 2
- Skin sensitiser Category 1B
- Specific target organ toxicity Category 3
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H341-Suspected of causing genetic defects.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Methylglyoxal
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'