Läs mer
Methylmalonyl DL-Carnitine Chloride (Mixture of Diastereomers), TRC
Produktkod. 30010894
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010894 50 mg
30136570 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30010894 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM31890050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 821794-54-1
Molekylformel C11H20ClNO6
Molekylvikt (g/mol) 297.73
Synonym 3-Carboxy-2-(2-carboxy-1-oxopropoxy)-N,N,N-trimethyl-1-propanaminium Chloride;
IUPAC-namn [3-carboxy-2-(2-carboxypropanoyloxy)propyl]-trimethylazanium;chloride
LEDER CC(C(=O)O)C(=O)OC(CC(=O)O)C[N+](C)(C)C.[Cl-]
Specifikationer
Kemiskt namn eller material Methylmalonyl DL-Carnitine Chloride (Mixture of Diastereomers)
Kvantitet 50 mg
InChI formel InChI=1S/C11H19NO6.ClH/c1-7(10(15)16)11(17)18-8(5-9(13)14)6-12(2,3)4;/h7-8H,5-6H2,1-4H3,(H-,13,14,15,16);1H
Formel vikt 297.0979