Läs mer
Methylparaoxon (Technical Grade), TRC
Produktkod. 30179645
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179645 1 g
30179182 100 mg
30179593 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179645 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3185601G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 950-35-6
Molekylformel C8 H10 N O6 P
Molekylvikt (g/mol) 247.14
Synonym Phosphoric acid, dimethyl p-nitrophenyl ester (6CI,7CI,8CI), Phenol, p-nitro-, dimethyl phosphate (6CI), 4-Nitrophenyl dimethylphosphate, BAY 11678, Desmethylnitrophos, Dimethyl 4-nitrophenyl phosphate, Dimethyl p-nitrophenyl phosphate, Dimethyl paraoxon, Methyl-E 600, Methylparaoxon, Methylparathion oxon, Paraoxon methyl, p-Nitrophenyl dimethyl phosphate
IUPAC-namn dimethyl (4-nitrophenyl) phosphate
LEDER COP(=O)(OC)Oc1ccc(cc1)[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material Paraoxon-methyl
Kvantitet 1 g
InChI formel InChI=1S/C8H10NO6P/c1-13-16(12,14-2)15-8-5-3-7(4-6-8)9(10)11/h3-6H,1-2H3
Formel vikt 247.0246